Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MFSLMASCC GWFKRWREPV RLANKQDKEG ALGEADVIEC LSLEKLVNEH KCLCQIEPCS AISGYGKKID KSIKKGLYWL LHVIARDFDA LNERIQKETT EQRALEEQEK QERAERVRKL REERKQNEQE QAELDGTSGL AELDPEPTNP FQPIASVIIE NEGKLEREKK NQKMEKDSDG CHLKHKMEHE QIETQGQVNH NGQKNNEFGL VENYKEALTQ QLKNEDETDR PSLESANGKK KTKKLRMKRN HRVEPLNIDD CAPESPTPPP PPPPVGWGTP KVTRLPKLEP LGETHHNDFY RKPLPPLAVP QRPNSDAHDV IS (1-321 aa encoded by BC094725)
This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintenance of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP ribosylation factor like GTPase 13B; ADP-ribosylation factor-like 2-like 1; ARL2-like protein 1; protein hennin; unnamed protein product
Gene Aliases: A530097K21Rik; A930014M17Rik; ARL2L1; C530009C10Rik; hnn; JBTS8
UniProt ID: (Human) Q3SXY8, (Mouse) Q640N2
Entrez Gene ID: (Dog) 487934, (Pig) 100624385, (Human) 200894, (Mouse) 68146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support