Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
ASH1 (ASH1L) is a member of the trithorax group of transcriptional activators. The protein contains four AT hooks, a SET domain, a PHD-finger motif, and a bromodomain. It is localized to many small speckles in the nucleus, and also to cell-cell tight junctions. Diseases associated with ASH1 include Mental Retardation, Autosomal Dominant 52 and Pulpitis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Absent small and homeotic disks protein 1 homolog; absent, small, or homeotic discs 1; ash1 (absent, small, or homeotic)-like; ASH1-like protein; chromatin remodeling factor; Histone-lysine N-methyltransferase ASH1L; huASH1; Lysine N-methyltransferase 2H; probable histone-lysine N-methyltransferase ASH1L
Gene Aliases: 8030453L17Rik; ASH1; ASH1L; ASH1L1; E430018P19Rik; KIAA1420; KMT2H
UniProt ID: (Human) Q9NR48, (Mouse) Q99MY8
Entrez Gene ID: (Human) 55870, (Mouse) 192195
Molecular Function: chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support