Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KAKEGHLPE KDGHGHEGRA AGEEAKQLAR VPSPYVRTPV VESARPNSTS SREAEPRKGE PAYENPKKSS EVKVKEERKE DHDLPPEAPQ THRASEPPPP NSSSSVHPGP LASMPMTVGV TGIHPMNSIS SLDRTRMMTP FMGISPLPGG ERFPYPSFHW DPIRDPLRDP YRELDIHRRD PLGRDFLLRN DPLHRLSTPR LYEADRSFRD REPHDYSHHH HHHHHPLSVD PRREHERGGH LDERERLHML REDYEHTRLH SVHPASLDGH LPHPSLITPG LPSMHYPRIS PTAGNQNGLL NKTPPTAALS APPPLISTLG GRPVSPRRTT PLSAEIRERP PSHTLKDIEA R (886-1235 aa encoded by BC064693)
AUTS2 also named as FBRSL2 or autism susceptibility candidate 2 is a 1259 amino acid protein, which belongs to the AUTS2 family. AUTS2 is highly expressed in brain, kidney and skeletal muscle. AUTS2 exists as four isoforms produced by alternative splicing events and is encoded by a gene located on human chromosome 7. The molecular weight of AUTS2 isoform is 65-70 kDa in the brain. The functional role of AUTS2 is not well known, although some studies have identified a putative role in transcriptional regulation during neuronal development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autism susceptibility gene 2 protein; autism-related protein 1
Gene Aliases: AUTS2; KIAA0442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support