Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MRPRRPLVFM SLVCALLNTC QAHRVHDDKP NIVLIMVDDL GIGDLGCYGN DTMRTPHIDR LAREGVRLTQ HISAASLCSP SRSAFLTGRY PIRSGMVSSG NRRVIQNLAV PAGLPLNETT LAALLKKQGY STGLIGKWHQ GLNCDSRSDQ CHHPYNYGFD YYYGMPFTLV DSCWPDPSRN TELAFESQLW LCVQLVAIAI; Positive Samples: 293T, A-549, 5637, 22Rv1, BT-474, Mouse liver, Mouse pancreas
Arylsulfatase F, also known as ARSF, is a 590 amino acid secretory protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Arylsulfatase F uses calcium as a cofactor to catalyze reactions that are important in maintaining correct bone composition. The activity of Arylsulfatase F, unlike that of other family members, such as Arylsulfatase E, is not inhibited by warfarin. The gene encoding Arylsulfatase F maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arylsulfatase F; ASF
Gene Aliases: ARSF; ASF
UniProt ID: (Human) P54793
Entrez Gene ID: (Human) 416
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support