Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLKLKNVFL AYFLVSIAGL LYALVQLGQP CDCLPPLRAA AEQLRQKDLR ISQLQAELRR PPPAPAQPPE PEALPTIYVV TPTYARLVQK AELVRLSQTL SLVPRLHWLL VEDAEGPTPL VSGLLAASGL LFTHLVVLTP KAQRLREGEP GWVHPRGVEQ RNKALDWLRG RGGAVGGEKD PPPPGTQGVV YFADDDNTYS RELFEEMRWT RGVSVWPVGL VGGLRFEGPQ VQDGRVVGFH TAWEPSRPFP VDMAGFAVAL PLLLDKPNAQ FDSTAPRGHL ESSLLSHLVD PKDLEPRAAN CTRVLVWHTR TEKPKMKQEE QLQRQGRGSD PAIEV
The protein encoded by this gene is a member of the glucuronyltransferase gene family, enzymes that exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product catalyzes the formation of the glycosaminoglycan-protein linkage by way of a glucuronyl transfer reaction in the final step of the biosynthesis of the linkage region of proteoglycans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-glucuronyltransferase 3; beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I); Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; GlcAT-I; GlcUAT-I; Glucuronosyltransferase I; Sqv-8-like protein; UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase
Gene Aliases: B3GAT3; GLCATI; glcUAT-I; JDSCD
UniProt ID: (Human) O94766
Entrez Gene ID: (Human) 26229
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support