Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDCSNGCSAE CTGEGGSKEV VGTFKAKDLI VTPATILKEK PDPNNLVFGT VFTDHMLTVE WSSEFGWEKP HIKPLQNLSL HPGSSALHYA VELFEGLKAF RGVDNKIRLF QPNLNMDRMY RSAVRATLPV FDKEELLECI QQLVKLDQEW VPYSTSASLY IRPTFIGTEP SLGVKKPTKA LLFVLLSPVG PYFSSGTFNP VSLWANPKYV RAWKGGTGDC KMGGNYGSSL FAQCEAVDNG CQQVLWLYGE DHQITEVGTM NLFLYWINED GEEELATPPL DGIILPGVTR RCILDLAHQW DTELSLFSIN LPDFLQFIYF
This gene encodes the cytosolic form of the enzyme branched-chain amino acid transaminase. This enzyme catalyzes the reversible transamination of branched-chain alpha-keto acids to branched-chain L-amino acids essential for cell growth. Two different clinical disorders have been attributed to a defect of branched-chain amino acid transamination: hypervalinemia and hyperleucine-isoleucinemia. As there is also a gene encoding a mitochondrial form of this enzyme, mutations in either gene may contribute to these disorders. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCAT(c); branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; Branched-chain-amino-acid aminotransferase, cytosolic; placental protein 18; Protein ECA39
Gene Aliases: BCAT1; BCATC; BCT1; ECA39; MECA39; PNAS121; PP18
UniProt ID: (Human) P54687
Entrez Gene ID: (Human) 586
Molecular Function: transaminase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support