Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAVAIAAARV WRLNRGLSQA ALLLLRQPGA RGLARSHPPR QQQQFSSLDD KPQFPGASAE FIDKLEFIQP NVISGIPIYR VMDRQGQIIN PSEDPHLPKE KVLKLYKSMT LLNTMDRILY ESQRQGRISF YMTNYGEEGT HVGSAAALDN TDLVFGQYRE AGVLMYRDYP LELFMAQCYG NISDLGKGRQ MPVHYGCKER HFVTISSPLA TQIPQAVGAA YAAKRANANR VVICYFGEGA ASEGDAHAGF NFAATLECPI IFFCRNNGYA ISTPTSEQYR GDGIAARGPG YGIMSIRVDG NDVFAVYNAT KEARRRAVAE NQPFLIEAMT YRIGHHSTSD DSSAYRSVDE VNYWDKQDHP ISRLRHYLLS QGWWDEEQEK AWRKQSRRKV MEAFEQAERK PKPNPNLLFS DVYQEMPAQL RKQQESLARH LQTYGEHYPL DHFDK
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid (BCAA) dehydrogenase complex (BCKD; EC 1.2.4.4), an inner-mitochondrial enzyme complex that catalyzes the oxidative decarboxylation of the branched-chain alpha-ketoacids derived from isoleucine, leucine, and valine. This reaction is the second major step in the catabolism of the branched-chain amino acids (Wynn et al., 1998). The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2), and a homodimeric dihydrolipoamide dehydrogenase (E3). E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoisovalerate dehydrogenase (lipoamide); 2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial; BCKDE1A; BCKDH E1-alpha; branched chain keto acid dehydrogenase E1 alpha protein; Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain
Gene Aliases: BCKDE1A; BCKDHA; MSU; MSUD1; OVD1A
UniProt ID: (Human) P12694
Entrez Gene ID: (Human) 593
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support