Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
The antibody reacts with the 37 kDa BMI1 protein.
Immunogen sequence: MHRTTRIKI TELNPHLMCV LCGGYFIDAT TIIECLHSFC KTCIVRYLET SKYCPICDVQ VHKTRPLLNI RSDKTLQDIV YKLVPGLFKN EMKRRRDFYA AHPSADAANG SNEDRGEVAD EDKRIITDDE IISLSIEFFD QNRLDRKVNK DKEKSKEEVN DKRYLRCPAA MTVMHLRKFL RSKMDIPNTF QIDVMYEEEP LKDYYTLMDI AYIYTWRRNG PLPLKYRVRP TCKRMKISHQ RDGLTNAGEL ESDSGSDKAN SPAGGIPSTS SCLPSPSTPV QSPHPQFPHI SSTMNGTSNS PSGNHQSSFA NRPRKSSVNG SSATSSG (1-326 aa encoded by BC011652)
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; Bmi1 polycomb ring finger protot-oncogene; COMMD3; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4 antibody; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; protein-binding protein; BMI1; putative; ring finger protein 51; RP11-573G6.1; unnamed protein product
Gene Aliases: Bmi-1; flvi-2/bmi-1; FLVI2/BMI1; PCGF4; RNF51
UniProt ID: (Human) P35226, (Mouse) P25916
Entrez Gene ID: (Human) 648, (Mouse) 12151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support