Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein similar to calcium/calmodulin dependent protein kinase, however, its exact function is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium/calmodulin-dependent protein kinase IG; Calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; CaM kinase IG; caM-KI gamma; CaMK-like CREB kinase III; caMKI gamma; caMKIG; CLICK III; RP1-272L16.2
Gene Aliases: CAMK1G; CLICK3; VWS1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support