Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Description: The monoclonal antibody X2/45-12 reacts with human CD124, which is also known as the IL-4R alpha chain. CD124 can associate with the common gamma chain to form the type I IL-4R or with IL-13Ralpha1 to form the type II IL-4R. While both receptor types can bind IL-4, only the type II receptor can associate with IL-13. The type I IL-4 receptor is found on T, B, NK, mast cells, basophils, and macrophages. The type II IL-4 receptor is found on many non-hematopoietic cells and macrophages. CD124 contains an ITIM that associates with and activates STAT6. IL-4 is responsible for the generation of high-affinity IgE antibodies and driving the polarization of naive Th0 cells into Th2 cells. In mucosal barriers, the cytokine is involved in the recruitment of innate and adaptive immune effector cells and mucous overproduction. Finally, IL-4R also exists as a soluble form that serves primarily as a carrier protein for IL-4 that increases its half-life.
The X2/45 antibody has been reported to have antagonist activity.
Applications Reported: This X2/45-12 antibody has been reported for use in flow cytometric analysis, and western blotting. (Please use Functional Grade purified X2/45-12, cat. 16-1249, in functional assays.).
Applications Tested: This X2/45-12 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD124; il-4 receptor; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; IL4R nirs variant 1; interleukin-4 receptor alpha chain; Interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha
Gene Aliases: 582J2.1; CD124; IL-4RA; IL4R; IL4RA
UniProt ID: (Human) P24394
Entrez Gene ID: (Human) 3566
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support