Product References
CD200-CD200R affects cisplatin and paclitaxel sensitivity by regulating cathepsin K-mediated p65 NF-κB signaling in cervical cancer.
Heliyon
Mou J,Zheng W,Wei D,Li D,Fan R,Tang Q
14-1639-82 was used in Western Blotting to investigate the role of CD200-CD200R in cervical cancer.
Tue Aug 01 00:00:00 EDT 2023
SARS-CoV-2 Delta (B.1.617.2) variant replicates and induces syncytia formation in human induced pluripotent stem cell-derived macrophages.
PeerJ
Thaweerattanasinp T,Wanitchang A,Saenboonrueng J,Srisutthisamphan K,Wanasen N,Sungsuwan S,Jongkaewwattana A,Chailangkarn T
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2023
A positive feedback loop: RAD18-YAP-TGF-β between triple-negative breast cancer and macrophages regulates cancer stemness and progression.
Cell death discovery
Yan X,He Y,Yang S,Zeng T,Hua Y,Bao S,Yang F,Duan N,Sun C,Liang Y,Fu Z,Huang X,Li W,Yin Y
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Tue Apr 12 00:00:00 EDT 2022
M2-like tumor-associated macrophages transmit exosomal miR-27b-3p and maintain glioblastoma stem-like cell properties.
Cell death discovery
Zhao G,Ding L,Yu H,Wang W,Wang H,Hu Y,Qin L,Deng G,Xie B,Li G,Qi L
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Thu Aug 04 00:00:00 EDT 2022
M1 macrophage-derived exosomes and their key molecule lncRNA HOTTIP suppress head and neck squamous cell carcinoma progression by upregulating the TLR5/NF-κB pathway.
Cell death & disease
Jiang H,Zhou L,Shen N,Ning X,Wu D,Jiang K,Huang X
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Thu Feb 24 00:00:00 EST 2022
TcpC Inhibits M1 but Promotes M2 Macrophage Polarization via Regulation of the MAPK/NF-κB and Akt/STAT6 Pathways in Urinary Tract Infection.
Cells
Fang J,Ou Q,Wu B,Li S,Wu M,Qiu J,Cen N,Hu K,Che Y,Ma Y,Pan J
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Sun Aug 28 00:00:00 EDT 2022
PTX3 mediates the infiltration, migration, and inflammation-resolving-polarization of macrophages in glioblastoma.
CNS neuroscience & therapeutics
Zhang H,Wang Y,Zhao Y,Liu T,Wang Z,Zhang N,Dai Z,Wu W,Cao H,Feng S,Zhang L,Cheng Q,Liu Z
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Pulmonary transplantation of alpha-1 antitrypsin (AAT)-transgenic macrophages provides a source of functional human AAT in vivo.
Gene therapy
Janosz E,Hetzel M,Spielmann H,Tumpara S,Rossdam C,Schwabbauer M,Kloos D,von Kaisenberg C,Schambach A,Buettner FFR,Janciauskiene S,Lachmann N,Moritz T
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Wed Sep 01 00:00:00 EDT 2021
Dysregulation of the CD163-Haptoglobin Axis in the Airways of COPD Patients.
Cells
Higham A,Baker JM,Jackson N,Shah R,Lea S,Singh D
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Tue Dec 21 00:00:00 EST 2021
Macrophages induce malignant traits in mammary epithelium via IKKε/TBK1 kinases and the serine biosynthesis pathway.
EMBO molecular medicine
Wilcz-Villega E,Carter E,Ironside A,Xu R,Mataloni I,Holdsworth J,Jones W,Moreno Béjar R,Uhlik L,Bentham RB,Godinho SA,Dalli J,Grose R,Szabadkai G,Jones L,Hodivala-Dilke K,Bianchi K
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Fri Feb 07 00:00:00 EST 2020
Extracorporeal Hemadsorption versus Glucocorticoids during Cardiopulmonary Bypass: A Prospective, Randomized, Controlled Trial.
Cardiovascular therapeutics
Taleska Stupica G,Sostaric M,Bozhinovska M,Rupert L,Bosnic Z,Jerin A,Ihan A,Klokocovnik T,Podbregar M
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2020
Patient iPSC-Derived Macrophages to Study Inborn Errors of the IFN-γ Responsive Pathway.
Cells
Haake K,Neehus AL,Buchegger T,Kühnel MP,Blank P,Philipp F,Oleaga-Quintas C,Schulz A,Grimley M,Goethe R,Jonigk D,Kalinke U,Boisson-Dupuis S,Casanova JL,Bustamante J,Lachmann N
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Wed Feb 19 00:00:00 EST 2020
CCL5 derived from tumor-associated macrophages promotes prostate cancer stem cells and metastasis via activating β-catenin/STAT3 signaling.
Cell death & disease
Huang R,Wang S,Wang N,Zheng Y,Zhou J,Yang B,Wang X,Zhang J,Guo L,Wang S,Chen Z,Wang Z,Xiang S
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Immunohistochemistry
Thu Apr 16 00:00:00 EDT 2020
Regional Activation of Myosin II in Cancer Cells Drives Tumor Progression via a Secretory Cross-Talk with the Immune Microenvironment.
Cell
Georgouli M,Herraiz C,Crosas-Molist E,Fanshawe B,Maiques O,Perdrix A,Pandya P,Rodriguez-Hernandez I,Ilieva KM,Cantelli G,Karagiannis P,Mele S,Lam H,Josephs DH,Matias-Guiu X,Marti RM,Nestle FO,Orgaz JL,Malanchi I,Fruhwirth GO,Karagiannis SN,Sanz-Moreno V
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Thu Feb 07 00:00:00 EST 2019
Recruitment of monocytes and epigenetic silencing of intratumoral CYP7B1 primarily contribute to the accumulation of 27-hydroxycholesterol in breast cancer.
American journal of cancer research
Shi SZ,Lee EJ,Lin YJ,Chen L,Zheng HY,He XQ,Peng JY,Noonepalle SK,Shull AY,Pei FC,Deng LB,Tian XL,Deng KY,Shi H,Xin HB
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
TIM‑4 blockade of KCs combined with exogenous TGF‑β injection helps to reverse acute rejection and prolong the survival rate of mice receiving liver allografts.
International journal of molecular medicine
Wu H,Xu X,Li J,Gong J,Li M
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
Biochanin a Enhances the Defense Against Salmonella enterica Infection Through AMPK/ULK1/mTOR-Mediated Autophagy and Extracellular Traps and Reversing SPI-1-Dependent Macrophage (MΦ) M2 Polarization.
Frontiers in cellular and infection microbiology
Zhao X,Tang X,Guo N,An Y,Chen X,Shi C,Wang C,Li Y,Li S,Xu H,Liu M,Wang Y,Yu L
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Fri Aug 30 00:00:00 EDT 2019
Similarities and differences between helminth parasites and cancer cell lines in shaping human monocytes: Insights into parallel mechanisms of immune evasion.
PLoS neglected tropical diseases
Narasimhan PB,Akabas L,Tariq S,Huda N,Bennuru S,Sabzevari H,Hofmeister R,Nutman TB,Tolouei Semnani R
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Widespread Virus Replication in Alveoli Drives Acute Respiratory Distress Syndrome in Aerosolized H5N1 Influenza Infection of Macaques.
Journal of immunology (Baltimore, Md. : 1950)
Wonderlich ER,Swan ZD,Bissel SJ,Hartman AL,Carney JP,O'Malley KJ,Obadan AO,Santos J,Walker R,Sturgeon TJ,Frye LJ,Maiello P,Scanga CA,Bowling JD,Bouwer AL,Duangkhae PA,Wiley CA,Flynn JL,Wang J,Cole KS,Perez DR,Reed DS,Barratt-Boyes SM
14163982 was used in immunohistochemistry to ask if small particle aerosols of virus would penetrate the lower respiratory tract and blanket alveoli where target cells reside
Wed Feb 15 00:00:00 EST 2017
Immortalization of primary microglia: a new platform to study HIV regulation in the central nervous system.
Journal of neurovirology
Garcia-Mesa Y,Jay TR,Checkley MA,Luttge B,Dobrowolski C,Valadkhan S,Landreth GE,Karn J,Alvarez-Carbonell D
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
Alternatively activated macrophages derived from THP-1 cells promote the fibrogenic activities of human dermal fibroblasts.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
Zhu Z,Ding J,Ma Z,Iwashina T,Tredget EE
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
MicroRNA-720 suppresses M2 macrophage polarization by targeting GATA3.
Bioscience reports
Zhong Y,Yi C
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Microenvironments in tuberculous granulomas are delineated by distinct populations of macrophage subsets and expression of nitric oxide synthase and arginase isoforms.
Journal of immunology (Baltimore, Md. : 1950)
Mattila JT,Ojo OO,Kepka-Lenhart D,Marino S,Kim JH,Eum SY,Via LE,Barry CE,Klein E,Kirschner DE,Morris SM,Lin PL,Flynn JL
14-1639 was used in Flow cytometry/Cell sorting to study macrophage diversity and function within the complex structures of granulomas.
Mon Jul 15 00:00:00 EDT 2013
Heat shock protein 27 differentiates tolerogenic macrophages that may support human breast cancer progression.
Cancer research
Banerjee S,Lin CF,Skinner KA,Schiffhauer LM,Peacock J,Hicks DG,Redmond EM,Morrow D,Huston A,Shayne M,Langstein HN,Miller-Graziano CL,Strickland J,O'Donoghue L,De AK
14-1639 was used in Flow cytometry/Cell sorting to investigate the role of Hsp27 in breast cancer, showing that Hsp27 differentiates tolerogenic macrophages.
Sat Jan 15 00:00:00 EST 2011
Interleukin-25 fails to activate STAT6 and induce alternatively activated macrophages.
Immunology
Stolfi C,Caruso R,Franzè E,Sarra M,De Nitto D,Rizzo A,Pallone F,Monteleone G
Published figure using CD163 monoclonal antibody (Product # 14-1639-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2011