Product References
Acetaminophen-Induced Liver Injury Alters the Acyl Ethanolamine-Based Anti-Inflammatory Signaling System in Liver.
Frontiers in pharmacology
Rivera P,Pastor A,Arrabal S,Decara J,Vargas A,Sánchez-Marín L,Pavón FJ,Serrano A,Bautista D,Boronat A,de la Torre R,Baixeras E,Lucena MI,de Fonseca FR,Suárez J
Published figure using CD45R monoclonal antibody (Product # RM2601) in Immunofluorescence
Thu Oct 01 00:00:00 EDT 2020
γδT cells but not αβT cells contribute to sepsis-induced white matter injury and motor abnormalities in mice.
Journal of neuroinflammation
Zhang X,Rocha-Ferreira E,Li T,Vontell R,Jabin D,Hua S,Zhou K,Nazmi A,Albertsson AM,Sobotka K,Ek J,Thornton C,Hagberg H,Mallard C,Leavenworth JW,Zhu C,Wang X
RM2601 was used in Flow cytometry/Cell sorting to study the contribution of different T cell subsets (αΒT cells and γδT cells) to white matter injury and consequent behavioural changes.
Wed Dec 20 00:00:00 EST 2017
Dysfunctional telomeres induce p53-dependent and independent apoptosis to compromise cellular proliferation and inhibit tumor formation.
Aging cell
Wang Y,Wang X,Flores ER,Yu J,Chang S
RM2601 was used in flow cytometry to elucidate induction of p53-dependent and independent apoptosis to compromise cellular proliferation and inhibition of tumor formation by dysfunctional telomeres
Mon Aug 01 00:00:00 EDT 2016
KSHV Latency Locus Cooperates with Myc to Drive Lymphoma in Mice.
PLoS pathogens
Sin SH,Kim Y,Eason A,Dittmer DP
RM2601 was used in flow cytometry to study lymphoma in mice driven by Myc cooperating with the KSHV latency locus
Tue Sep 01 00:00:00 EDT 2015
Targeting cell surface TLR7 for therapeutic intervention in autoimmune diseases.
Nature communications
Kanno A,Tanimura N,Ishizaki M,Ohko K,Motoi Y,Onji M,Fukui R,Shimozato T,Yamamoto K,Shibata T,Sano S,Sugahara-Tobinai A,Takai T,Ohto U,Shimizu T,Saitoh S,Miyake K
RM2601 was used in Flow cytometry/Cell sorting to show that Toll-like receptor 7 is present on the cell surface of immune cells and that TLR7 responses can be inhibited by an anti-TLR7 antibody.
Wed Feb 04 00:00:00 EST 2015
Conventional bone marrow-derived dendritic cells contribute to toll-like receptor-independent production of alpha/beta interferon in response to inactivated parapoxvirus ovis.
Journal of virology
Siegemund S,Hartl A,von Buttlar H,Dautel F,Raue R,Freudenberg MA,Fejer G,Büttner M,Köhler G,Kirschning CJ,Sparwasser T,Alber G
RM2601 was used in flow cytometry to study the effect of inactivated Parapoxvirus ovis on dendritic cells
Tue Sep 01 00:00:00 EDT 2009
Id3 restricts the developmental potential of gamma delta lineage during thymopoiesis.
Journal of immunology (Baltimore, Md. : 1950)
Ueda-Hayakawa I,Mahlios J,Zhuang Y
RM2601 was used in Flow cytometry/Cell sorting to show that functional loss of Id3 increases gammadelta T cell production from progenitor cells undergoing TCRbeta rearrangement.
Fri May 01 00:00:00 EDT 2009
B-lymphocyte depletion ameliorates Sjögren's syndrome in Id3 knockout mice.
Immunology
Hayakawa I,Tedder TF,Zhuang Y
RM2601 was used in flow cytometry to investigate the role of B cells in Sjogren's syndrome in a mouse model
Sat Sep 01 00:00:00 EDT 2007
IgE enhances specific antibody and T-cell responses in mice overexpressing CD23.
Scandinavian journal of immunology
Carlsson F,Hjelm F,Conrad DH,Heyman B
RM2601 was used in flow cytometry and immunohistochemistry to investigate if IgE enhances antibody and T-cell responses in IgE receptor CD23 transgenic animals or if CD23's downregulatory effect precludes IgE-mediated enhancement
Fri Sep 14 00:00:00 EDT 2007
Gut ischemia-reperfusion affects gut mucosal immunity: a possible mechanism for infectious complications after severe surgical insults.
Critical care medicine
Fukatsu K,Sakamoto S,Hara E,Ueno C,Maeshima Y,Matsumoto I,Mochizuki H,Hiraide H
RM2601 was used in flow cytometry to determine the influence of gut ischemia/reperfusion on gut-associated lymphoid tissue mass and function
Sun Jan 01 00:00:00 EST 2006
TLR4, TLR9 and MyD88 are not required for the positive selection of autoreactive B cells into the primary repertoire.
European journal of immunology
Silver K,Ferry H,Crockford T,Cornall RJ
RM2601 was used in flow cytometry to demonstrate that the effects of Toll-like receptors on autoimmunity occur in the established repertoire and not during B cell development
Thu Jun 01 00:00:00 EDT 2006
The peritoneal cavity provides a protective niche for B1 and conventional B lymphocytes during anti-CD20 immunotherapy in mice.
Journal of immunology (Baltimore, Md. : 1950)
Hamaguchi Y,Uchida J,Cain DW,Venturi GM,Poe JC,Haas KM,Tedder TF
RM2601 was used in flow cytometry and immunohistochemistry - frozen section to determine the effect of anti-CD20 immunotherapy on tissue B cells in vivo
Fri Apr 01 00:00:00 EST 2005
The earliest thymic progenitors in adults are restricted to T, NK, and dendritic cell lineage and have a potential to form more diverse TCRbeta chains than fetal progenitors.
Journal of immunology (Baltimore, Md. : 1950)
Lu M,Tayu R,Ikawa T,Masuda K,Matsumoto I,Mugishima H,Kawamoto H,Katsura Y
RM2601 was used in flow cytometry to study T cell progenitors in the adult thymus
Tue Nov 01 00:00:00 EST 2005
APS, an adaptor molecule containing PH and SH2 domains, has a negative regulatory role in B cell proliferation.
Biochemical and biophysical research communications
Iseki M,Kubo-Akashi C,Kwon SM,Yamaguchi A,Takatsu K,Takaki S
RM2601 was used in flow cytometry to use transgenic mice to study the functions of APS in B cell development and activation
Fri May 13 00:00:00 EDT 2005
Dendritic cell expansion occurs in mesenteric lymph nodes of B10.BR mice infected with the murine nematode parasite Trichuris muris.
Parasitology research
Koyama K
RM2601 was used in flow cytometry to characterize dendritic cells in mice infected with Trichuris muris
Sat Oct 01 00:00:00 EDT 2005
Hematopoietic stem cell dose correlates with the speed of immune reconstitution after stem cell transplantation.
Blood
Chen BJ,Cui X,Sempowski GD,Domen J,Chao NJ
RM2601 was used in flow cytometry to test if higher numbers of hematopoietic stem cells correlate with the speed of immune reconstitution in a congenic transplantation mouse model
Tue Jun 01 00:00:00 EDT 2004
Development and functional analysis of eosinophils from murine embryonic stem cells.
British journal of haematology
Hamaguchi-Tsuru E,Nobumoto A,Hirose N,Kataoka S,Fujikawa-Adachi K,Furuya M,Tominaga A
RM2601 was used in flow cytometry to generate and characterize eosinophils from murine embryonic stem cells.
Mon Mar 01 00:00:00 EST 2004
Reduced apoptosis and ameliorated listeriosis in TRAIL-null mice.
Journal of immunology (Baltimore, Md. : 1950)
Zheng SJ,Jiang J,Shen H,Chen YH
RM2601 was used in flow cytometry to identify a role for TRAIL in apoptosis during listeriosis
Mon Nov 01 00:00:00 EST 2004
Increased numbers of B-1 cells and enhanced responses against TI-2 antigen in mice lacking APS, an adaptor molecule containing PH and SH2 domains.
Molecular and cellular biology
Iseki M,Kubo C,Kwon SM,Yamaguchi A,Kataoka Y,Yoshida N,Takatsu K,Takaki S
RM2601 was used in flow cytometry to characterize adaptor molecule containing PH and SH2 domains knock out mice
Mon Mar 01 00:00:00 EST 2004
Extensive proliferation of T cell lineage-restricted progenitors in the thymus: an essential process for clonal expression of diverse T cell receptor beta chains.
European journal of immunology
Kawamoto H,Ohmura K,Fujimoto S,Lu M,Ikawa T,Katsura Y
RM2601 was used in flow cytometry to assess the proliferative potential of thymic progenitors that are T cell lineage restricted.
Sat Mar 01 00:00:00 EST 2003
Passive transfer of flt-3L-derived dendritic cells delays diabetes development in NOD mice and associates with early production of interleukin (IL)-4 and IL-10 in the spleen of recipient mice.
Clinical and experimental immunology
Morin J,Faideau B,Gagnerault MC,Lepault F,Boitard C,Boudaly S
RM2601 was used in flow cytometry to assess the contribution of DC function in non-obese diabetic mice.
Mon Dec 01 00:00:00 EST 2003
Graft-versus-leukemia effect after suicide-gene-mediated control of graft-versus-host disease.
Blood
Litvinova E,Maury S,Boyer O,Bruel S,Benard L,Boisserie G,Klatzmann D,Cohen JL
RM2601 was used in flow cytometry to investigate graft-versus-leukemia.
Sun Sep 15 00:00:00 EDT 2002
An analysis of T cell intrinsic roles of E2A by conditional gene disruption in the thymus.
Journal of immunology (Baltimore, Md. : 1950)
Pan L,Hanrahan J,Li J,Hale LP,Zhuang Y
RM2601 was used in flow cytometry to explore the role of E2A in T cell lineage.
Mon Apr 15 00:00:00 EDT 2002
Cell-intrinsic effects of non-MHC NOD genes on dendritic cell generation in vivo.
International immunology
Prasad SJ,Goodnow CC
RM2601 was used in flow cytometry to characterize the differences in dendritic cell generation in vivo caused by non-MHC NOD genes.
Sat Jun 01 00:00:00 EDT 2002
SH2-B is required for both male and female reproduction.
Molecular and cellular biology
Ohtsuka S,Takaki S,Iseki M,Miyoshi K,Nakagata N,Kataoka Y,Yoshida N,Takatsu K,Yoshimura A
RM2601 was used in Flow cytometry/Cell sorting to study the role of SH2-B in the insulin-like growth factor-mediated reproductive pathway in mice.
Wed May 01 00:00:00 EDT 2002
NK1.1+ cell depletion in vivo fails to prevent protection against infection with the murine nematode parasite Trichuris muris.
Parasite immunology
Koyama K
RM2601 was used in flow cytometry to test if NKT cells produce IL-4 in response to T. muris infection and induce an Th2 response.
Tue Jun 03 00:00:00 EDT 2003
Extrafollicular plasmablast B cells play a key role in carrying retroviral infection to peripheral organs.
Journal of immunology (Baltimore, Md. : 1950)
Finke D,Baribaud F,Diggelmann H,Acha-Orbea H
RM2601 was used in flow cytometry to explore the migration properties of B cells after differentiation in murine peripheral lymph node infected with mouse mammary tumor virus.
Tue May 15 00:00:00 EDT 2001
Immature multipotent hemopoietic progenitors lacking long-term bone marrow-reconstituting activity in the aorta-gonad-mesonephros region of murine day 10 fetuses.
Journal of immunology (Baltimore, Md. : 1950)
Ohmura K,Kawamoto H,Lu M,Ikawa T,Ozaki S,Nakao K,Katsura Y
RM2601 was used in flow cytometry to examine multipotent progenitors from early fetuses.
Thu Mar 01 00:00:00 EST 2001
Langerhans cells develop from a lymphoid-committed precursor.
Blood
Anjuère F,del Hoyo GM,Martín P,Ardavín C
RM2601 was used in flow cytometry to determine the lymphoid or myeloid origin of Langerhans cells.
Fri Sep 01 00:00:00 EDT 2000
Presence of B220 within thymocytes and its expression on the cell surface during apoptosis.
Immunology
Oka S,Mori N,Matsuyama S,Takamori Y,Kubo K
RM2601 was used in flow cytometry to test if B220 contributes to apoptosis of immature T cells.
Tue Aug 01 00:00:00 EDT 2000
Preservation of graft-versus-infection effects after suicide gene therapy for prevention of graft-versus-host disease.
Human gene therapy
Cohen JL,Saron MF,Boyer O,Thomas-Vaslin V,Bellier B,Lejeune L,Charlotte F,Klatzmann D
RM2601 was used in flow cytometry to assess the recovery of functional antiviral immune responses in ganciclovir-treated mice that express viral thymidine kinase
Sun Dec 10 00:00:00 EST 2000
Immunophenotypical and functional heterogeneity of dendritic cells generated from murine bone marrow cultured with different cytokine combinations: implications for anti-tumoral cell therapy.
Immunology
Masurier C,Pioche-Durieu C,Colombo BM,Lacave R,Lemoine FM,Klatzmann D,Guigon M
RM2601 was used in flow cytometry to characterize in vitro-generated dendritic cells.
Thu Apr 01 00:00:00 EST 1999
Cellular and molecular characterization of the scurfy mouse mutant.
Journal of immunology (Baltimore, Md. : 1950)
Clark LB,Appleby MW,Brunkow ME,Wilkinson JE,Ziegler SF,Ramsdell F
RM2601 was used in flow cytometry to further characterize mice hemizygous for scurfy.
Mon Mar 01 00:00:00 EST 1999
Emergence of T cell progenitors without B cell or myeloid differentiation potential at the earliest stage of hematopoiesis in the murine fetal liver.
Journal of immunology (Baltimore, Md. : 1950)
Kawamoto H,Ohmura K,Fujimoto S,Katsura Y
RM2601 was used in flow cytometry to assess if progenitors colonizing the thymus are T cell lineage restricted.
Mon Mar 01 00:00:00 EST 1999
Emergence of T, B, and myeloid lineage-committed as well as multipotent hemopoietic progenitors in the aorta-gonad-mesonephros region of day 10 fetuses of the mouse.
Journal of immunology (Baltimore, Md. : 1950)
Ohmura K,Kawamoto H,Fujimoto S,Ozaki S,Nakao K,Katsura Y
RM2601 was used in flow cytometry to investigate the aorta-gonad-mesonephros region as a source of hemopoietic progenitors.
Mon Nov 01 00:00:00 EST 1999
Impaired immune responses and B-cell proliferation in mice lacking the Id3 gene.
Molecular and cellular biology
Pan L,Sato S,Frederick JP,Sun XH,Zhuang Y
RM2601 was used in flow cytometry to investigate the role of Id3 in tissue and embryo development.
Wed Sep 01 00:00:00 EDT 1999
B220 is expressed on apoptotic thymocytes induced by X-irradiation.
The Journal of veterinary medical science
Oka S,Kubo K,Matsuyama S,Takamori Y
RM2601 was used in flow cytometry to determine the expression of B220 on murine thymocytes after whole-body X-irradiation.
Thu Apr 01 00:00:00 EST 1999
Viral superantigen-induced negative selection of TCR transgenic CD4+ CD8+ thymocytes depends on activation, but not proliferation.
Blood
Ferrero I,Anjuère F,Azcoitia I,Renno T,MacDonald HR,Ardavín C
RM2601 was used in flow cytometry to analyze the cell activation and proliferation requirements for the deletion of viral superantigen-reactive Vbeta8.1 T-cell receptor transgenic thymocytes.
Mon Jun 01 00:00:00 EDT 1998
Repression of B7.2 on self-reactive B cells is essential to prevent proliferation and allow Fas-mediated deletion by CD4(+) T cells.
The Journal of experimental medicine
Rathmell JC,Fournier S,Weintraub BC,Allison JP,Goodnow CC
RM2601 was used in flow cytometry to report that B7.2 repression is critical to eliminate autoreactive B cells by Fas.
Mon Aug 17 00:00:00 EDT 1998
Three populations of mouse lymph node dendritic cells with different origins and dynamics.
Journal of immunology (Baltimore, Md. : 1950)
Salomon B,Cohen JL,Masurier C,Klatzmann D
RM2601 was used in flow cytometry to characterize different populations of mouse lymph node dendritic cells.
Thu Jan 15 00:00:00 EST 1998
IL-6-deficient mice are resistant to experimental autoimmune encephalomyelitis: roles of IL-6 in the activation and differentiation of autoreactive T cells.
Journal of immunology (Baltimore, Md. : 1950)
Samoilova EB,Horton JL,Hilliard B,Liu TS,Chen Y
RM2601 was used in flow cytometry to assess experimental autoimmune encephalomyelitis in mice deficient in IL-6.
Tue Dec 15 00:00:00 EST 1998
Self-reactive B cells are not eliminated or inactivated by autoantigen expressed on thyroid epithelial cells.
The Journal of experimental medicine
Akkaraju S,Canaan K,Goodnow CC
RM2601 was used in flow cytometry to investigate how humoral tolerance is maintained in the presence of organ-specific membrane antigens.
Mon Dec 15 00:00:00 EST 1997
Isolation of the most immature population of murine fetal thymocytes that includes progenitors capable of generating T, B, and myeloid cells.
The Journal of experimental medicine
Hattori N,Kawamoto H,Katsura Y
RM2601 was used in flow cytometry to report that FcR is a useful marker to isolate the most immature population of murine fetal thymic cells.
Fri Nov 01 00:00:00 EST 1996
Impaired antigen specific responses and enhanced polyclonal stimulation in mice infected with Ehrlichia muris.
Microbiology and immunology
Kawahara M,Suto C,Shibata S,Futohashi M,Rikihisa Y
RM2601 was used in flow cytometry to characterize the immune responses of BALB/c mice infected with Ehrlichia muris.
Tue Mar 25 00:00:00 EST 1997
Reduced development of CD4-8-B220+ T cells but normal autoantibody production in lpr/lpr mice lacking major histocompatibility complex class I molecules.
European journal of immunology
Ohteki T,Iwamoto M,Izui S,MacDonald HR
RM2601 was used in flow cytometry to examine the origin of double negative T cells using Ipr mice.
Sun Jan 01 00:00:00 EST 1995
CD3+ and CD4+ cells adoptively transfer experimental hypersensitivity pneumonitis.
The American review of respiratory disease
Schuyler M,Gott K,Shopp G,Crooks L
RM2601 was used in flow cytometry to describe the cells that transfer adoptive murine experimental hypersensitivity pneumonitis.
Tue Dec 01 00:00:00 EST 1992