Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD318; CUB domain-containing protein 1; gp140; Membrane glycoprotein gp140; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; Transmembrane and associated with src kinases
Gene Aliases: 9030022E12Rik; AA409659; CD318; CDCP1; E030027H19Rik; SIMA135; TRASK; UNQ2486/PRO5773
UniProt ID: (Human) Q9H5V8, (Mouse) Q5U462
Entrez Gene ID: (Human) 64866, (Mouse) 109332
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support