Product References
The APC/C activator Cdh1 regulates the G2/M transition during differentiation of placental trophoblast stem cells.
Biochemical and biophysical research communications
Naoe H,Chiyoda T,Ishizawa J,Masuda K,Saya H,Kuninaka S
MA5-12981 was used in western blot to study the role of Cdh1 in regulating G2/M transition during placental differentiation
Fri Jan 11 00:00:00 EST 2013
Menthol inhibits the proliferation and motility of prostate cancer DU145 cells.
Pathology oncology research : POR
Wang Y,Wang X,Yang Z,Zhu G,Chen D,Meng Z
MA5-12981 was used in western blot to study the effects of TRPM8 activation on prostate cancer cell proliferation and migration
Mon Oct 01 00:00:00 EDT 2012
Cyclic AMP inhibits the proliferation of thyroid carcinoma cell lines through regulation of CDK4 phosphorylation.
Molecular biology of the cell
Rocha AS,Paternot S,Coulonval K,Dumont JE,Soares P,Roger PP
MA5-12981 was used in immunoprecipitation to study the role of CDK4 phosphorylation in the mechanism by which cAMP inhibits thyroid carcinoma cell proliferation
Sat Nov 01 00:00:00 EDT 2008
Regulation of keratinocyte proliferation in rats with deep, partial-thickness scald: modulation of cyclin D1-cyclin-dependent kinase 4 and histone H1 kinase activity of M-phase promoting factor.
The Journal of surgical research
Xie T,Niu Y,Ge K,Lu S
MA5-12981 was used in western blot to study the mechanism of regulation of keratinocyte proliferation in rats with deep partial-thickness scald
Sun Jun 01 00:00:00 EDT 2008
Differential modification of p27Kip1 controls its cyclin D-cdk4 inhibitory activity.
Molecular and cellular biology
James MK,Ray A,Leznova D,Blain SW
MA5-12981 was used in immunoprecipitation to study if the growth state-dependent tyrosine phosphorylation of p27Kip1 determines whether or not it inhibits cdk4
Tue Jan 01 00:00:00 EST 2008
Inhibition of prostate cancer growth by muscadine grape skin extract and resveratrol through distinct mechanisms.
Cancer research
Hudson TS,Hartle DK,Hursting SD,Nunez NP,Wang TT,Young HA,Arany P,Green JE
MA5-12981 was used in western blot to study the distinct mechanisms by which muscadine grape skin extract and resveratrol inhibit prostate cancer cell growth
Sat Sep 01 00:00:00 EDT 2007
The LxCxE pRb interaction domain of cyclin D1 is dispensable for murine development.
Cancer research
Landis MW,Brown NE,Baker GL,Shifrin A,Das M,Geng Y,Sicinski P,Hinds PW
MA5-12981 was used in western blot to study whether the LxCxE domain of cyclin D1 which interacts with the retinoblastoma protein is required for normal murine development
Wed Aug 15 00:00:00 EDT 2007
Critical requirement for cell cycle inhibitors in sustaining nonproliferative states.
The Journal of cell biology
Pajalunga D,Mazzola A,Salzano AM,Biferi MG,De Luca G,Crescenzi M
MA5-12981 was used in immunoprecipitation to examine the role of cell cycle inhibitors in cell growth arrest
Mon Mar 12 00:00:00 EDT 2007
The role of p28GANK in rat oval cells activation and proliferation.
Liver international : official journal of the International Association for the Study of the Liver
Shan YF,Zhou WP,Fu XY,Yan HX,Yang W,Liu SQ,Cao HF,Kang B,Wu MC,Wang HY
MA5-12981 was used in western blot to investigate the importance of p28GANK in the activation and proliferation of rat oval cells
Wed Mar 01 00:00:00 EST 2006
Requirement for CDK4 kinase function in breast cancer.
Cancer cell
Yu Q,Sicinska E,Geng Y,Ahnström M,Zagozdzon A,Kong Y,Gardner H,Kiyokawa H,Harris LN,Stål O,Sicinski P
MA5-12981 was used in immunoprecipitation to study the importance of cyclin D1-dependent Cdk4 kinase activity in mammary tumorigenesis
Sun Jan 01 00:00:00 EST 2006
In vitro and in vivo pharmacokinetic-pharmacodynamic relationships for the trisubstituted aminopurine cyclin-dependent kinase inhibitors olomoucine, bohemine and CYC202.
Clinical cancer research : an official journal of the American Association for Cancer Research
Raynaud FI,Whittaker SR,Fischer PM,McClue S,Walton MI,Barrie SE,Garrett MD,Rogers P,Clarke SJ,Kelland LR,Valenti M,Brunton L,Eccles S,Lane DP,Workman P
MA5-12981 was used in western blot to study the pharmacokinetics and pharmacodynamics of three tri-substituted aminopurine Cdk inhibitors in a model of colon carcinoma
Fri Jul 01 00:00:00 EDT 2005
p21Cip1 and p27Kip1 induce distinct cell cycle effects and differentiation programs in myeloid leukemia cells.
The Journal of biological chemistry
Muñoz-Alonso MJ,Acosta JC,Richard C,Delgado MD,Sedivy J,León J
MA5-12981 was used in immunoprecipitation to study the role of p21Cip1 and p27Kip1 in the cell cycle and differentiation programs of myeloid leukemia cells
Fri May 06 00:00:00 EDT 2005
p107 inhibits G1 to S phase progression by down-regulating expression of the F-box protein Skp2.
The Journal of cell biology
Rodier G,Makris C,Coulombe P,Scime A,Nakayama K,Nakayama KI,Meloche S
MA5-12981 was used in western blot to study the role of Skp2 in the mechanism by which p107 inhibits G1/S phase progression
Mon Jan 03 00:00:00 EST 2005
p53, p16 and cyclin D1: molecular determinants of radiotherapy treatment response in oral carcinoma.
International journal of cancer
Jayasurya R,Francis G,Kannan S,Lekshminarayanan K,Nalinakumari KR,Abraham T,Abraham EK,Nair MK
MA5-12981 was used in immunohistochemistry to study the molecular determinants of the response to radiotherapy in oral carcinoma
Sat May 01 00:00:00 EDT 2004
Regulation and role of p21 and p27 cyclin-dependent kinase inhibitors during hepatocyte differentiation and growth.
American journal of physiology. Gastrointestinal and liver physiology
Ilyin GP,Glaise D,Gilot D,Baffet G,Guguen-Guillouzo C
MA5-12981 was used in western blot to study the role of the p21 and p27 cyclin-dependent kinase inhibitors in hepatocyte development and proliferation
Tue Jul 01 00:00:00 EDT 2003
Interaction of Hsp90 with the nascent form of the mutant epidermal growth factor receptor EGFRvIII.
The Journal of biological chemistry
Lavictoire SJ,Parolin DA,Klimowicz AC,Kelly JF,Lorimer IA
MA5-12981 was used in western blot to study the interaction of nascent EGFRvIII with Hsp90 and its importance for maintaining EGFRvIII expression
Fri Feb 14 00:00:00 EST 2003
Autoimmune responses to proliferating cell nuclear antigen multiprotein complexes involved in cell proliferation are strongly associated with their structure and biologic function in patients with systemic lupus erythematosus.
Arthritis and rheumatism
Kogure T,Takasaki Y,Takeuchi K,Yamada H,Nawata M,Ikeda K,Matsushita M,Matsudaira R,Kaneda K,Terasawa K,Hashimoto H
MA5-12981 was used in ELISA to study the mechanism of autoimmune responses to PCNA and its associated proteins in systemic lupus erythematosus
Fri Nov 01 00:00:00 EST 2002
Targeted disruption of CDK4 delays cell cycle entry with enhanced p27(Kip1) activity.
Molecular and cellular biology
Tsutsui T,Hesabi B,Moons DS,Pandolfi PP,Hansel KS,Koff A,Kiyokawa H
MA5-12981 was used in western blot to study the role of CDK4 in cell cycle regulation using targeted knockdown
Fri Oct 01 00:00:00 EDT 1999