Search Thermo Fisher Scientific
MA1-90203 detects CDw78 from rat samples.
MA1-90203 has been successfully used in immunohistochemistry (frozen tissue) applications.
The MA1-90203 immunogen is: L-CA (CD45)
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.