Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: GHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ AAAQARCTAR GGSLAQPADR QQMEALTRYL RAALAPYNWP VWLGVHDRRA EGLYLFENGQ RVSFFAWHRS PRPELGAQPS ASPHPLSPDQ PNGGTLENCV AQASDDGSWW DHDCQRRLYY VCEFPF.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human SCGF-alpha. Anti-Human SCGF-alpha-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect Human SCGF-alpha by sandwich ELISA (using 100 µL/well) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human SCGF-alpha (500-P162) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human SCGF-alpha.
Western Blot: To detect Human SCGF-alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human SCGF-alpha is 1.5-3.0 ng/lane, under either reducingor non-reducing conditions.
500-P162BT-1 mg will be provided as 2 x 500 µg
This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b SCGF; beta SCGF; betaSCGF; bSCGF; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; Lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; p47; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; Stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF
Gene Aliases: CLEC11A; CLECSF3; LSLCL; P47; SCGF
UniProt ID: (Human) Q9Y240
Entrez Gene ID: (Human) 6320
Molecular Function: intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support