Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRHSKRTYCP DWDDKDWDYG KWRSSSSHKR RKRSHSSAQE NKRCKYNHSK MCDSHYLESR SINEKDYHSR RYIDEYRNDY TQGCEPGHRR RDHESRYQNH SSKSSGRSGR SSYKSKHRIH HSTSHRRSHG KSHRRKRTRS VEDDEEGRLI CQSGDVLSAR YEIVDTLGEG AFGKVVECID HKAGGRHVAV KIVKNVDRYC EAARSEIQVL EHLNTTDPNS TFRCVQMLEW FEHHGHICIV FELLGLSTYD FIKENGFLPF RLDHIRKMAY QICKSVNFLH SNKLTHTDLK PENILFVQSD YTEAYNPKIK RDERTLINPD IKVVDFGSAT YDDEHHSTLV STRHYRAPEV ILALGWSQPC DVWSIGCILI EYYLGFTVFP THDSKEHLAM MERILGPLPK HMIQKTRKRK YFHHDRLDWD EHSSAGRYVS RRCKPLKEFM LSQDVEHERL FDLIQKMLEY DPAKRITLRE ALKHPFFDLL KKSI
CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing. Therefore, the encoded protein may play an indirect role in governing splice site selection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC-like kinase 1; CDC28/CDC2-like kinase; Dual specificity protein kinase CLK1; Protein kinase CLK1; protein tyrosine kinase STY
Gene Aliases: CLK; CLK/STY; CLK1; STY
UniProt ID: (Human) P49759
Entrez Gene ID: (Human) 1195
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support