Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MVRTDGHTLS EKRNYQVTNS MFGASRKKFV EGVDSDYHDE NMYYSQSSMF PHRSEKDMLA SPSTSGQLSQ FGASLYGQQS ALGLPMRGMS NNTPQLNRSL SQGTQLPSHV TPTTGVPTMS LHTPPSPSRG ILPMNPRNMM NHSQVGQGIG IPSRTNSMSS SGLGSPNRSS PSIICMPKQQ PSRQPFTVNS MSGFGMNRNQ AFGMNNSLSS NIFNGTDGSE NVTGLDLSDF PALADRNRRE GSGNPTPLIN PLAGRAPYVG MVTKPANEQS QDFSIHNEDF PALPGSSYKD PTSSNDDSKS NLNTSGKTTS STDGPKFPGD KSSTTQNNNQ QKKGIQVLPD GRVTNIPQGM VTDQFGMIGL LTFIRAAETD PGMVHLALGS DLTTLGLNLN SPENLYPKFA SPWASSPCRP QDIDFHVPSE YLTNIHIRDK LAAIKLGRYG EDLLFYLYYM NGGDVLQLLA AVELFNRDWR YHKEERVWIT RAPGMEPTMK TNTYERGTYY FFDCLNWRKV AKEFHLEYDK LEERPHLPST FNYNPAQQAF
CCR4-NOT Transcription Complex 2 (CNOT2) is a component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. CNOT2 is required for the CCR4-NOT complex structural integrity. It can repress transcription and may link the CCR4-NOT complex to transcriptional regulation; the repressive function may specifically involve the N-Cor repressor complex containing HDAC3, NCOR1 and NCOR2. CNOT2 is involved in the maintenance of embryonic stem (ES) cell identity. (UniProt)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCR4-associated factor 2; CCR4-NOT transcription complex subunit 2; CCR4-NOT transcription complex, subunit 2; negative regulator of transcription 2
Gene Aliases: CDC36; CNOT2; HSPC131; MSTP046; NOT2; NOT2H
UniProt ID: (Human) Q9NZN8
Entrez Gene ID: (Human) 4848
Molecular Function: general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support