Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: ELEKVVLKST FNRLGGLQFD KELRSLIAYL TTVTTWTIRD KFARLSQMAT ILNLERVTEI LDYWGPNSGP LTWRLTPAEV RQVLALRIDF RSEDIKRLRL
Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COG complex subunit 4; complexed with Dor1p; Component of oligomeric Golgi complex 4; conserved oligomeric Golgi complex protein 4; Conserved oligomeric Golgi complex subunit 4
Gene Aliases: CDG2J; COD1; COG4
UniProt ID: (Human) Q9H9E3
Entrez Gene ID: (Human) 25839
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support