Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin-binding; Clabp coronin; Clipin-A; coronin, actin binding protein, 1A; coronin-1; Coronin-1A; Coronin-like protein A; Coronin-like protein p57; FLJ41407; MGC117380; TACO; Tryptophan aspartate-containing coat protein
Gene Aliases: CLABP; CLIPINA; CORO1; CORO1A; HCORO1; IMD8; p57; TACO
UniProt ID: (Human) P31146
Entrez Gene ID: (Human) 11151
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support