Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CRTAC1 encodes a glycosylated extracellular matrix protein that is found in the interterritorial matrix of articular deep zone cartilage. This protein is used as a marker to distinguish chondrocytes from osteoblasts and mesenchymal stem cells in culture. The presence of FG-GAP motifs and an RGD integrin-binding motif suggests that this protein may be involved in cell-cell or cell-matrix interactions. Copy number alterations in this gene have been observed in neurofibromatosis type 1-associated glomus tumors. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; Cartilage acidic protein 1; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; FLJ10320
Gene Aliases: ASPIC; ASPIC1; CEP-68; CEP68; CRTAC1
UniProt ID: (Human) Q9NQ79
Entrez Gene ID: (Human) 55118
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support