Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: AENRLAQHTL QALQSELDSL RADNIKLFEK IKFLQSYPGR GSGSDDTELR YSSQYEERLD PFSSFSKRER QRKYLSLSPW DKATLSMGRL VLSNKMARTI
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCAAT displacement protein; CDP; CDP/Cux p200; cut homolog; cut-like homeobox 1; golgi integral membrane protein 6; Homeobox protein cut-like 1; Homeobox protein cux-1; Protein CASP; putative protein product of Nbla10317
Gene Aliases: CASP; CDP; CDP/Cut; CDP1; Clox; COY1; CUTL1; CUX; Cux/CDP; CUX1; GOLIM6; Nbla10317; p100; p110; p200; p75
UniProt ID: (Human) Q13948
Entrez Gene ID: (Human) 1523
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support