Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: U-87MG, U-251MG, MCF7, Mouse liver, Mouse brain, Rat liver. The target is usually found in the following locations: Endoplasmic reticulum membrane, Microsome membrane, Single-pass membrane protein.
Immunogen sequence: HRARSRYEHI PGPPRPSFLL GHLPCFWKKD EVGGRVLQDV FLDWAKKYGP VVRVNVFHKT SVIVTSPESV KKFLMSTKYN KDSKMYRALQ TVFGERLFGQ GLVSECNYER WHKQRRVIDL AFSRSSLVSL METFNEKAEQ LVEILEAKAD GQTPVSMQDM LTYTAMDILA KAAFGMETSM LLGAQKPLSQ AVKLMLEGIT ASRNTLAKFL PGKRKQLREV RESIRFLRQV GRDWVQRRRE ALKRGEEVPA DILTQIL
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is expressed in the brain, where it converts cholesterol to 24S-hydroxycholesterol. While cholesterol cannot pass the blood-brain barrier, 24S-hydroxycholesterol can be secreted in the brain into the circulation to be returned to the liver for catabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CH24H; Cholesterol 24-hydroxylase; Cholesterol 24-monooxygenase; Cholesterol 24S-hydroxylase; cholestrol 24-hydroxylase; Cytochrome P450 46A1; cytochrome P450, 46 (cholesterol 24-hydroxylase); cytochrome P450, 46a1 (cholesterol 24-hydroxylase); cytochrome P450, family 46, subfamily A, polypeptide 1; cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase)
Gene Aliases: CP46; CYP46; CYP46A1
UniProt ID: (Human) Q8N2B0, (Mouse) Q9WVK8
Entrez Gene ID: (Human) 10858, (Rat) 362782, (Mouse) 13116
Molecular Function: oxidoreductase oxygenase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support