Search Thermo Fisher Scientific
MA1-22685 detects parvovirus in virus samples.
MA1-22685 has been successfully used in ELISA procedures.
The MA1-22685 immunogen is canine parvovirus.
Canine parvovirus is a contagious virus mainly affecting dogs. CPV is highly contagious and is spread from dog to dog by direct or indirect contact with their feces. Vaccines can prevent this infection, but mortality can reach 91% in untreated cases. Treatment often involves veterinary hospitalization.
Protein Aliases: CPV; parvo