Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Collagen X Antibodies

          Invitrogen

          Collagen X Monoclonal Antibody (X53), eBioscience™

          8 Published Figures
          23 References
          View all (10) Collagen X antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Collagen X Monoclonal Antibody (X53), eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (8)
          Collagen X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Collagen X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Collagen X Antibody (14-9771-82) in IHC (P)

          Immunohistochemistry of formalin-fixed paraffin embedded human infiltrating ductal carcinoma tissue using 10 µg/mL of Mouse IgG1 K Isotype Control Purified (left) or 10 µg/mL of Anti-Collagen Type X Purified (right) followed by Anti-Mouse IgG Biotin, Streptavidin HRP, and DAB visualization.Nuclei are counterstained with hematoxylin. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Collagen X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Collagen X Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed) (IHC, IHC (PFA))
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Western Blot (WB)

          Product Details

          14-9771-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 18 publications 18 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          10 µg/mL
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Dog, Guinea pig, Human, Mouse, Pig, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          X53

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • eFluor 570
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2573018

          Product Specific Information

          Description: The monoclonal antibody X53 recognizes human, mouse, porcine, guinea pig, and rat Collagen Type X. X53 does not cross-react with collagen types I, II, III, IV, or VI. Collagen Type X is a homotrimeric short chain collagen. Collagen Type X has limited expression in normal adult tissues, primarily being expressed during bone growth and development. Expression is localized to the peri- and extracellular matrix of hypertrophic chondrocytes during endochondral ossification and also plays a role in cartilage mineralization. Collagen Type X is elevated in many tumor types with expression being localized in the vasculature. Mutations to the gene COL10A1, which encodes human Collagen Type X, is associated with Schmid type metaphyseal chondrodysplasia.

          Applications Reported: This X53 antibody has been reported for use in microscopy, immunohistochemical staining, ELISA, and immunocytochemistry.

          Applications Tested: This X53 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Collagen alpha-1(X) chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1

          View more View less

          Gene Aliases: Col10; Col10a-1; COL10A1

          View more View less

          UniProt ID: (Human) Q03692, (Mouse) Q05306

          View more View less

          Entrez Gene ID: (Pig) 448809, (Human) 1300, (Dog) 100856711, (Mouse) 12813, (Rat) 25681, (Guinea pig) 106026924

          View more View less

          Function(s)
          protein binding metal ion binding molecular_function extracellular matrix structural constituent extracellular matrix structural protein
          Process(es)
          skeletal system development endochondral ossification extracellular matrix organization collagen catabolic process cartilage development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-56756dfb8b-7vhdw:80/100.66.79.199:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0