Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: STQLSNLGST ETLEEMPSGS QDKSAPSGHK HLTVEELFGT SLPKEQPAVV GLDSEEMERL PGDASQKEPN SFLPFPFEQL GGAPQSETLG VPSAAHHSVQ
Signal transduction of TGF-beta superfamily members is regulated by Smad proteins. In particular, Smads influence specific gene transcription by relaying signals from the cell membrane to the nucleus. Smad4 plays an essential role in TGF-beta-induced transcriptional activation wherein phosphorylated receptor-associated Smads associate with Smad4. Furthermore, SMIF (Smad4-intereacting protein) and Smad4 complex with TGF-beta and BMP4. An increase in Smad4 concentration increases the translocation of this complex to the nucleus. SMIF and Smad4 interact directly through a EVH1/WH1 domain on SMIF and a proline-rich activation domain on Smad4. Smad4 is essential to nuclear translocation of SMIF as deletion of the Smad4-interacting domain (located in the N-terminal 100 amino acids) of SMIF eliminates TGF-beta-induced nuclear translocation of SMIF. The human SMIF gene is ubiquitously expressed and encodes a protein with a relative molecular mass of 70 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCP1 decapping enzyme homolog A; DCP1 decapping enzyme-like protein A; decapping enzyme hDcp1a; FLJ21691; mRNA-decapping enzyme 1A; putative protein product of Nbla00360; Smad4-interacting transcriptional co-activator; Transcription factor SMIF
Gene Aliases: DCP1A; HSA275986; Nbla00360; SMAD4IP1; SMIF
UniProt ID: (Human) Q9NPI6
Entrez Gene ID: (Human) 55802
Molecular Function: mRNA capping factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support