Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Humn BD-4. Anti-Human BD-4-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect hBD-4 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BD-4 (500-P268) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBD-4.
Western Blot: To detect hBD-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBD-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P268BT-1MG will be provided as 2 x 500 µg
Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BD-4; Beta-defensin 104; Beta-defensin 4; Defensin, beta 104; defensin, beta 104A; defensin, beta 4
Gene Aliases: BD-4; DEFB-4; DEFB104; DEFB104A; DEFB104B; DEFB4; hBD-4
UniProt ID: (Human) Q8WTQ1
Entrez Gene ID: (Human) 140596
Molecular Function: antimicrobial response protein defense/immunity protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support