Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MPVRKQDTQR ALHLLEEYRS KLSQTEDRQL RSSIERVINI FQSNLFQALI DIQEFYEVTL LDNPKCIDRS KPSEPIQPVN TWEISSLPSS
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs, large homolog 1; discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; Disks large homolog 1; DKFZp761P0818; DKFZp781B0426; hDlg; presynaptic protein SAP97; SAP-97; Synapse-associated protein 97
Gene Aliases: dJ1061C18.1.1; DLG1; DLGH1; hdlg; SAP-97; SAP97
UniProt ID: (Human) Q12959
Entrez Gene ID: (Human) 1739
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support