Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GNLHVTQGAG RFIPCSPFSD YVYKRIKARR KMADLHAVPR GMYDGPVFDL TTTPKGGTPA GSARGSPTRP NPPVRNLHQS GFSLSGTQVD EGVRSASKR
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collapsin response mediator protein 4; collapsin response mediator protein 4 long; CRMP-4; dihydropyrimidinase-like 3; Dihydropyrimidinase-related protein 3; DRP-3; testicular secretory protein Li 7; tuc-4; ULIP-1; Unc-33-like phosphoprotein 1
Gene Aliases: CRMP-4; CRMP4; DPYSL3; DRP-3; DRP3; LCRMP; ULIP; ULIP-1; ULIP1
UniProt ID: (Human) Q14195
Entrez Gene ID: (Human) 1809
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support