Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with Monkey, Cat, Pig, Sheep and Bovine.
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
DR3/TNFRSF25 is a member of the TNF-receptor superfamily. This receptor is expressed preferentially in the tissues enriched in lymphocytes, and it may play a role in regulating lymphocyte homeostasis. This receptor has been shown to stimulate NF-kappa B activity and regulate cell apoptosis. The signal transduction of this receptor is mediated by various death domain containing adaptor proteins. Knockout studies in mice suggested the role of DR3/TNFRSF25 in the removal of self-reactive T cells in the thymus. Multiple alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported, most of which are potentially secreted molecules. The alternative splicing of this gene in B and T cells encounters a programmed change upon T-cell activation, which predominantly produces full-length, membrane bound isoforms, and is thought to be involved in controlling lymphocyte proliferation induced by T-cell activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-3; apoptosis inducing receptor; Apoptosis-inducing receptor AIR; Apoptosis-mediating receptor DR3; Apoptosis-mediating receptor TRAMP; death domain receptor 3 soluble form; Death receptor 3; death receptor beta; LARD; Lymphocyte-associated receptor of death; Protein WSL; Protein WSL-1; RP4-650H14.2; Tumor necrosis factor receptor superfamily member 25; tumor necrosis factor receptor superfamily, member 12; tumor necrosis factor receptor superfamily, member 12 (translocating chain-association membrane protein); tumor necrosis factor receptor superfamily, member 25
Gene Aliases: APO-3; APO3; DDR3; DR3; LARD; TNFRSF12; TNFRSF25; TR3; TRAMP; UNQ455/PRO779; WSL; WSL-1; WSL-LR; WSL1
UniProt ID: (Human) Q93038
Entrez Gene ID: (Human) 8718, (Rat) 500592, (Mouse) 85030
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support