Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200-1:500 | View 1 publication 1 publication |
Immunohistochemistry (Frozen) (IHC (F)) |
1:1,000 | - |
Immunocytochemistry (ICC/IF) |
1:200-1:1,000 | - |
Immunoprecipitation (IP) |
- | View 1 publication 1 publication |
Miscellaneous PubMed (Misc) |
- | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Mouse, Rat |
Published species |
Marsupial |
Host/Isotype |
Mouse / IgG2b, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
SG2-D1a |
Immunogen |
Recombinant rat D1a dopamine receptor (123 residues with 23 residues near the C-terminus).[Swiss-Prot# P18901] |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
tris glycine with 0.15M NaCl |
Contains |
0.05% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2261891 |
This antibody does not react with human, canine, porcine, sheep, rabbit, or bovine.
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D(1A) dopamine receptor; D1 receptor; D1A dopamine receptor; D1a receptor; Dopamine 1 receptor; Dopamine D1 receptor; dopamine receptor D1A; Dopamine-1A receptor; G protein-coupled receptor DRD1
Gene Aliases: C030036C15Rik; D1a; Drd-1; Drd1; Drd1a; Gpcr15
UniProt ID: (Mouse) Q61616
Entrez Gene ID: (Mouse) 13488, (Rat) 24316
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support