Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LARTYHTQAR HLTLDPASKS LGLPHPGAPA AASTATLPQR TLAMPAPPAG TAPPAPGPTP AEPPTAPSAA PPAPSTEPPR AGGPHTKMGG SRDSLLEMST
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
DSCAML1 encodes a protein that is a member of the Ig superfamily of cell adhesion molecules and is involved in neuronal differentiation. The encoded membrane-bound protein localizes to the cell surface, where it forms aggregates that repel neuronal processes of the same cell type.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support