Product References
Anti-cancer binary system activated by bacteriophage HK022 integrase.
Oncotarget
Elias A,Gritsenko N,Gorovits R,Spector I,Prag G,Yagil E,Kolot M
MA5-14344 was used in Immunohistochemistry-immunofluorescence to study the specificity towards cancer cells of the HK022 integrase based binary system, and compare this to the mono system.
Fri Jun 08 00:00:00 EDT 2018
Repair of oxidative DNA damage, cell-cycle regulation and neuronal death may influence the clinical manifestation of Alzheimer's disease.
PloS one
Silva AR,Santos AC,Farfel JM,Grinberg LT,Ferretti RE,Campos AH,Cunha IW,Begnami MD,Rocha RM,Carraro DM,de Bragança Pereira CA,Jacob-Filho W,Brentani H
MA5-14344 was used in immunohistochemistry - paraffin section to study the clinical manifestation of Alzheimer's disease involving cell-cycle regulation, neuronal death and repair of oxidative DNA damage
Thu Feb 12 00:00:00 EST 2015
The relationship between E2F family members and tumor growth in colorectal adenocarcinomas: A comparative immunohistochemical study of 100 cases.
Applied immunohistochemistry & molecular morphology : AIMM
Xanthoulis A,Kotsinas A,Tiniakos D,Fiska A,Tentes AA,Kyroudi A,Kittas C,Gorgoulis V
MA5-14344 was used in immunohistochemistry to study the relationship between the immunohistochemical expression of E2F1, E2F2 and E2F4 and colorectal adenocarcinoma growth
Tue Jul 01 00:00:00 EDT 2014
Identification of prognostic biomarkers for glioblastomas using protein expression profiling.
International journal of oncology
Jung Y,Joo KM,Seong DH,Choi YL,Kong DS,Kim Y,Kim MH,Jin J,Suh YL,Seol HJ,Shin CS,Lee JI,Kim JH,Song SY,Nam DH
MA5-14344 was used in immunohistochemistry - paraffin section to identify diagnostic and prognostic markers for glioblastoma
Sun Apr 01 00:00:00 EDT 2012
CDK4/6 inhibition antagonizes the cytotoxic response to anthracycline therapy.
Cell cycle (Georgetown, Tex.)
McClendon AK,Dean JL,Rivadeneira DB,Yu JE,Reed CA,Gao E,Farber JL,Force T,Koch WJ,Knudsen ES
MA5-14344 was used in western blot to study the protective effect of pharmacological inhibition of CDK4/6 against antracycline cytotoxicity in triple-negative breast cancer
Sun Jul 15 00:00:00 EDT 2012
Impact of E2F-1 expression on clinical outcome of gastric adenocarcinoma patients with adjuvant chemoradiation therapy.
Clinical cancer research : an official journal of the American Association for Cancer Research
Lee J,Park CK,Park JO,Lim T,Park YS,Lim HY,Lee I,Sohn TS,Noh JH,Heo JS,Kim S,Lim DH,Kim KM,Kang WK
MA5-14344 was used in immunohistochemistry - paraffin section to assess expression and prognostic significance of E2F-1 and thymidylate synthase in R(0)-resected gastric adenocarcinoma patients
Tue Jan 01 00:00:00 EST 2008
Enhanced expression of 70-kilodalton heat shock protein limits cell division in a sepsis-induced model of acute respiratory distress syndrome.
Critical care medicine
Bromberg Z,Raj N,Goloubinoff P,Deutschman CS,Weiss YG
MA5-14344 was used in western blot to examine the effect of exogenous 70-kilodalton heat shock protein expression on the pathophysiology of acute respiratory distress syndrome
Tue Jan 01 00:00:00 EST 2008
Deregulated expression of p16INK4a and p53 pathway members in benign and malignant myoepithelial tumours of the salivary glands.
Histopathology
Vékony H,Röser K,Löning T,Raaphorst FM,Leemans CR,Van der Waal I,Bloemena E
MA5-14344 was used in immunohistochemistry to study the expression of p16INK4a and p53 pathway members in benign and malignant myoepithelial salivary gland tumours
Mon Dec 01 00:00:00 EST 2008
Overexpression of N-Myc rapidly causes acute myeloid leukemia in mice.
Cancer research
Kawagoe H,Kandilci A,Kranenburg TA,Grosveld GC
MA5-14344 was used in western blot to determine whether N-Myc overexpression is highly oncogenic in mouse myeloid cells
Thu Nov 15 00:00:00 EST 2007
Evaluation of immunohistochemical markers in non-small cell lung cancer by unsupervised hierarchical clustering analysis: a tissue microarray study of 284 cases and 18 markers.
The Journal of pathology
Au NH,Cheang M,Huntsman DG,Yorida E,Coldman A,Elliott WM,Bebb G,Flint J,English J,Gilks CB,Grimes HL
MA5-14344 was used in immunohistochemistry to evaluate immunohistochemical markers of non-small cell lung cancer using unsupervised hierarchical clustering analysis
Wed Sep 01 00:00:00 EDT 2004
UCN-01 suppresses E2F-1 mediated by ubiquitin-proteasome-dependent degradation.
Clinical cancer research : an official journal of the American Association for Cancer Research
Hsueh CT,Wu YC,Schwartz GK
MA5-14344 was used in western blot to investigate the mechniasm for the suppression of E2F-1 expression by UCN-01
Thu Mar 01 00:00:00 EST 2001