Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive test controls include: HeLa, NIH/3T3, Jurkat, Mouse liver, Rat spleen. The target is usually found in the following locations: Cytoplasm, Nucleus.
Immunogen sequence: IYGVLNRKRG HVFEESQVAG TPMFVVKAYL PVNESFGFTA DLRSNTGGQA FPQCVFDHWQ ILPGDPFDNS SRPSQVVAET RKRKGLKEGI PALDNFLDKL
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-2; Elongation factor 2; elongation factor-2; polypeptidyl-tRNA translocase
Gene Aliases: EEF-2; EEF2; EF-2; EF2; SCA26
UniProt ID: (Human) Q58J86, (Mouse) P58252, (Rat) P05197
Entrez Gene ID: (Human) 1938, (Mouse) 13629, (Rat) 29565
Molecular Function: translation elongation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support