Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Positive test controls include: HeLa, THP-1, U-251MG, Mouse liver, Mouse brain, Mouse spleen, Rat lung, Rat testis, Rat kidney. The target is usually found in the following locations: Cell membrane, Cell projection, Cytoplasmic side, Early endosome membrane, Peripheral membrane protein, Recycling endosome membrane, cilium membrane.
Immunogen sequence: MFSWVSKDAR RKKEPELFQT VAEGLRQLYA QKLLPLEEHY RFHEFHSPAL EDADFDNKPM VLLVGQYSTG KTTFIRHLIE QDFPGMRIGP EPTTDSFIAV
EHD1 acts in early endocytic membrane fusion and membrane trafficking of recycling endosomes. EHD1 also plays a role in myoblast fusion and is recruited to endosomal membranes upon nerve growth factor stimulation, indirectly regulates neurite outgrowth. Further, EHD1 belongs to a highly conserved gene family encoding EPS15 homology (EH) domain-containing proteins. The protein-binding EH domain was first noted in EPS15, a substrate for the epidermal growth factor receptor. The EH domain has been shown to be an important motif in proteins involved in protein-protein interactions and in intracellular sorting. The EHD1 protein is thought to play a role in the endocytosis of IGF1 receptors. Alternatively spliced transcript variants have been found for the EHD1 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain-containing protein 1; EH-domain containing 1; hPAST1; mPAST1; PAST homolog 1; Testilin
Gene Aliases: AA409636; CDABP0131; EHD1; H-PAST; HPAST1; PAST; PAST1; RGD1306960; RME-1
UniProt ID: (Human) Q9H4M9, (Mouse) Q9WVK4, (Rat) Q641Z6
Entrez Gene ID: (Human) 10938, (Mouse) 13660, (Rat) 293692
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support