Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DGQEYAQVIK MLGNGRLEAM CFDGVKRLCH IRGKLRKKVW INTSDIILVG LRDYQDNKAD VILKYNADEA RSLKAYGELP EHAKINETDT
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-1A Y isoform; eIF-4C; eukaryotic translation initiation factor 1A, Y chromosome; Eukaryotic translation initiation factor 1A, Y-chromosomal; Eukaryotic translation initiation factor 4C
Gene Aliases: eIF-4C; EIF1AY
UniProt ID: (Human) O14602
Entrez Gene ID: (Human) 9086
Molecular Function: translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support