Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 8
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSASQDSRSR DNGPDGMEPE GVIESNWNEI VDSFDDMNLS ESLLRGIYAY GFEKPSAIQQ RAILPCIKGY DVIAQAQSGT GKTATFAISI LQQIELDLKA
The eIF4A proteins are involved in many aspects of RNA metabolism due to their RNA-binding capacity and ATPase activity. eIF4A1 and eIF4A2 are cytoplasmic, whereas eIF4A3 is nuclear-localized. In the current model of translation initiation, eIF4A unwinds RNA secondary structures in the 5'-UTR of mRNAs which is necessary to allow efficient binding of the small ribosomal subunit, and subsequent scanning for the initiator codon.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-1; ATP-dependent RNA helicase eIF4A-2; ATP-dependent RNA helicase eIF4A-3; DDX48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; eIF-4A-I; eIF-4A-II; eIF-4A-III; eIF4A-I; eIF4A-II; eukaryotic initiation factor 4A-like NUK-34; eukaryotic initiation factor 4AI; eukaryotic translation initiation factor 4A; initiation factor eIF-4A long form; KIAA0111; NMP 265; nuclear matrix protein 265; protein synthesis initiation factor 4AII; unnamed protein product
Gene Aliases: 2400003O03Rik; 4833432N07Rik; BM-010; DDX2A; DDX2B; DDX48; EIF-4A; eIF-4A-I; eIF-4A-II; eIF-4A-III; Eif4; EIF4A; eIF4A-I; eIF4A-II; eIF4A-III; eIF4AIII; EIF4F; Fal1; mKIAA0111; MUK34; NEDHSS; NMP265; NUK34; RCPS
UniProt ID: (Human) P60842, (Human) Q14240, (Human) P38919, (Mouse) P60843, (Mouse) P10630, (Rat) Q5RKI1, (Mouse) Q91VC3, (Rat) Q3B8Q2
Entrez Gene ID: (Human) 1973, (Human) 1974, (Human) 9775, (Rat) 287436, (Mouse) 13681, (Mouse) 13682, (Rat) 303831, (Mouse) 192170, (Rat) 688288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support