Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: DVAVLKARAK LLQKYLCDEQ KELQALYALQ ALVVTLEQPP NLLRMFFDAL YDEDVVKEDA FYSWESSKDP AEQQGKGVAL KSVTAFFKWL REAEEESDHN
The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686A1451; eIF-4-gamma 1; EIF4-gamma; eucaryotic translation initiation factor 4G; Eukaryotic translation initiation factor 4 gamma 1; eukaryotic translation initiation factor 4 gamma, 1; OTTHUMP00000210577; OTTHUMP00000210586; OTTHUMP00000210587; OTTHUMP00000210588; OTTHUMP00000210595; OTTHUMP00000210602; OTTHUMP00000210603; p220
Gene Aliases: E030015G23Rik; eIF-4-gamma 1; eIF-4G 1; EIF-4G1; EIF4F; EIF4G; EIF4G1; EIF4GI; P220; PARK18
UniProt ID: (Human) O43177, (Mouse) Q6NZJ6
Entrez Gene ID: (Human) 1981, (Mouse) 208643, (Rat) 287986
Molecular Function: translation factor translation initiation factor translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support