Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGLTAAAPR PGVLLLLLSI LHPSRPGGVP GAIPGGVPGG VFYPGAGLGA LGGGALGPGG KPLKPVPGGL AGAGLGAGLG AFPAVTFPGA LVPGGVADAA AAYKAAKAGA GLGGVPGVGG LGVSAGAVVP QPGAGVKPGK VPGVGLPGVY PGGVLPGARF PGVGVLPGVP TGAGVKPKAP GVGGAFAGIP GVGPFGGPQP GVPLGYPIKA PKLPGYGPGG VAGAAGKAGY PTGTGVGPQA AAAAAAKAAA KFGAGAAGVL PGVGGAGVPG VPGAIPGIGG IAGVGTPAAA AAAAAAAKAA KYGAAAGLVP GGPGFGPGVV GVPGAGVPGV GVPGAGIPVV PGAGIPGAAV PGVVSPEAAA KAAAKAAKYG ARPGVGVGGI PTYGVGAGGF PGFGVGVGGI PGVAGVPSVG GVPGVGGVPG VGISPEAQAA AAAKAAKYGL VPGVGVAPGV GVAPGVGVAP GVGLAPGVGV APGVGVAPGV GVAPGIGPGG VAGAAAGLGA GIPGLGVGVG VPGLGVGAGV PGLGVGAGVP GFRAVPGALA AAKAAKYGAA VPGVLGGLGA LGGVGIPGGV VGAGPAAAAA AAKAAAKAAQ FGLVGAAGLG GLGVGGLGVP GVGGLGGIPP AAAAKAAKYG VAARPGFGLS PIFPGGACLG KACGRKRK
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Elastin; FLJ38671; FLJ43523; Tropoelastin
Gene Aliases: ELN; SVAS; WBS; WS
UniProt ID: (Human) P15502
Entrez Gene ID: (Human) 2006
Molecular Function: structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support