Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: TKLQANTNLW NEYRIKDLGQ FYAGLSAESY PIIKGVACKV ASLFDSNQIC EKAFSYLTRN QHTLSQPLTD EHLQALFRVA TTEMEPGWDD LVRERNESN
The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EPM2A (laforin) interacting protein 1; EPM2A-interacting protein 1; Laforin-interacting protein
Gene Aliases: A930003G21Rik; EPM2AIP1; KIAA0766; mKIAA0766; My007
UniProt ID: (Human) Q7L775, (Mouse) Q8VEH5
Entrez Gene ID: (Human) 9852, (Mouse) 77781, (Rat) 316021
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support