Invitrogen
MA1-10658 detects Entamoeba histolytica.
MA1-10658 has been successfully used in ELISA applications.
The MA1-10658 immunogen is Entamoeba histolytica HK-9.
Entamoeba histolytica is an anaerobic parasitic amoebozoan, part of the genus Entamoeba. Predominantly infecting humans and other primates causing amoebiasis, E. histolytica is estimated to infect about 50 million people worldwide. E. histolytica infection is estimated to kill more than 55,000 people each year.
Protein Aliases: E. histolytica