Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
FAM192A is also known as Nip30. This protein has not bee characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 192, member A; NEFA-interacting nuclear protein NIP30; PA28G-interacting protein; protein FAM192A; PSME3-interacting protein
Gene Aliases: 1700001O11Rik; 2310065K24Rik; AI596259; C16orf94; CDA018; CDA10; FAM192A; NIP30; PIP30; PSME3IP1
UniProt ID: (Human) Q9GZU8, (Mouse) Q91WE2
Entrez Gene ID: (Human) 80011, (Mouse) 102122
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support