Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA5-16899 targets Fli-1 in WB applications and shows reactivity with Human, mouse, and Rat samples.
The PA5-16899 immunogen is synthetic peptide derived from C-terminus of human Fli-1.
ETS-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes ETS-1, ETS-2, ERG-1, ERG-2, ELK, E74, FLI-1, PU. 1 and PEA3. Members of the ETS gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localization signal sequence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ewing sarcoma breakpoint region 2; Friend leukemia integration 1 transcription factor; Friend leukemia virus integration 1; Proto-oncogene Fli-1; Retroviral integration site protein Fli-1; Transcription factor ERGB
Gene Aliases: Fli-1; FLI1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support