Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HVFRGRPGSG FSSTSLGHLG PKCEPHYTGG ETEVQNKGLE PVSRQWQRLR PFDLGRAHWS PIQGGVVDLH RRGSPVCRPG PTLKGLCYPS GIEAATAQGR WGQHAVPSGL
Plays a role in cardiac repolarization possibly by stabilizing membrane expression of the potassium channel KCNH2/HERG, or by assisting its synthesis, folding or export from the endoplasmic reticulum, in a heat shock protein-dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein 207
Gene Aliases: C1orf188; RNF207
UniProt ID: (Human) Q6ZRF8
Entrez Gene ID: (Human) 388591
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support