Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with Monkey, Horse, Cat
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Checkpoint suppressor 1; CHES; Forkhead box protein N3
Gene Aliases: C14orf116; CHES1; FOXN3; PRO1635
UniProt ID: (Human) O00409
Entrez Gene ID: (Human) 1112
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support