Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RSKPYTRRPK PPYSYIALIA MAIRDSAGGR LTLAEINEYL MGKFPFFRGS YTGWRNSVRH NLSLNDCFVK VLRDPSRPWG KDNYWMLNPN SEYTFADGVF RRRRKRLSHR
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead box protein Q1; Hepatocyte nuclear factor 3 forkhead homolog 1; HFH-1; HNF-3/forkhead-like protein 1; winged helix/forkhead transcription factor
Gene Aliases: FOXQ1; HFH1
UniProt ID: (Human) Q9C009
Entrez Gene ID: (Human) 94234
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support