Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MADYSTVPPP SSGSAGGGGG GGGGGGVNDA FKDALQRARQ IAAKIGGDAG TSLNSNDYGY GGQKRPLEDG DQPDAKKVAP QNDSFGTQLP PMHQQQRSVM TEEYKVPDGM VGFIIGRGGE QISRIQQESG CKIQIAPDSG GLPERSCMLT GTPESVQSAK RLLDQIVEKG RPAPGFHHGD GPGNAVQEIM IPASKAGLVI GKGGETIKQL QERAGVKMVM IQDGPQNTGA DKPLRITGDP YKVQQAKEMV LELIRDQGGF REVRNEYGSR IGGNEGIDVP IPRFAVGIVI GRNGEMIKKI QNDAGVRIQF KPDDGTTPER IAQITGPPDR CQHAAEIITD LLRSVQAGNP GGPGPGGRGR GRGQGNWNMG PPGGLQEFNF IVPTGKTGLI IGKGGETIKS ISQQSGARIE LQRNPPPNAD PNMKLFTIRG TPQQIDYARQ LIEEKIGGPV NPLGPPVPHG PHGVPGPHGP PGPPGPGTPM GPYNPAPYNP GPPGPAPHGP PAPYAPQGWG NAYPHWQQQA PPDPAKAGTD PNSAAWAAYY AHYYQQQAQP PPAAPAGAPT TTQTNGQGDQ QNPAPAGQVD YTKAWEEYYK KMGQAVPAPT GAPPGGQPDY SAAWAEYYRQ QAAYYAQTSP QGMPQHPPAP QCRFDPASIE LAL
This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA helicase V; far upstream element (FUSE) binding protein 1; Far upstream element-binding protein 1; FBP; FUSE binding protein 1; hDH V
Gene Aliases: FBP; FUBP; FUBP1; hDH V
UniProt ID: (Human) Q96AE4
Entrez Gene ID: (Human) 8880
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support