Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-32744 detects GPRv40 from human samples.
GPRv40 is an Orphan-A GPCR with no known ligand. ESTs have been isolated from human B-cell/lung/testis, brain, parathyroid, testis, and fetus/fibroblast/ovary/parathyroid/placenta libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GPRv40; Probable G-protein coupled receptor 150; seven transmembrane helix receptor
Gene Aliases: GPR150; PGR11
UniProt ID: (Human) Q8NGU9
Entrez Gene ID: (Human) 285601
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support