Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALRGVSVRL LSRGPGLHVL RTWVSSAAQT EKGGRTQSQL AKSSRPEFDW QDPLVLEEQL TTDEILIRDT FRTYCQERLM PRILLANRNE VFHREIISEM GELGVLGPTI KGYGCAGVSS VAYGLLAREL ERVDSGYRSA MSVQSSLVMH PIYAYGSEEQ RQKYLPQLAK GELLGCFGLT EPNSGSDPSS METRAHYNSS NKSYTLNGTK TWITNSPMAD LFVVWARCED GCIRGFLLEK GMRGLSAPRI QGKFSLRASA TGMIIMDGVE VPEENVLPGA SSLGGPFGCL NNARYGIAWG VLGASEFCLH TARQYALDRM QFGVPLARNQ LIQKKLADML TEITLGLHAC LQLGRLKDQD KAAPEMVSLL KRNNCGKALD IARQARDMLG GNGISDEYHV IRHAMNLEAV NTYEGTHDIH ALILGRAITG IQAFTASK
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family. It catalyzes the oxidative decarboxylation of glutaryl-CoA to crotonyl-CoA and CO(2) in the degradative pathway of L-lysine, L-hydroxylysine, and L-tryptophan metabolism. It uses electron transfer flavoprotein as its electron acceptor. The enzyme exists in the mitochondrial matrix as a homotetramer of 45-kD subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCD; Glutaryl-CoA dehydrogenase, mitochondrial; glutaryl-Coenzyme A dehydrogenase
Gene Aliases: ACAD5; GCD; GCDH
UniProt ID: (Human) Q92947
Entrez Gene ID: (Human) 2639
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support