Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: TAHLSVVAED GSAVSATSTI NLYFGSKVRS PVSGILFNNE MDDFSSPSIT NEFGVPPSPA NFIQPGKQPL SSMCPTIMVG QDGQVRMVVG
Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD224; Gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGT 1; glutathione hydrolase 1; Glutathione hydrolase 1 proenzyme; Leukotriene-C4 hydrolase; testicular tissue protein Li 73
Gene Aliases: CD224; D22S672; D22S732; dwg; GGT; GGT 1; GGT-1; GGT1; Ggtp; GTG
UniProt ID: (Human) P19440, (Mouse) Q60928
Entrez Gene ID: (Human) 2678, (Mouse) 14598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support